Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002133.2 | NP_002124.1 | 2 | 196 | ||
GenBank mRNA | AK313120.1 | - | 2 | 166 | ||
BC001491.2 | - | 2 | 155 | 3504480 | ||
BP288767.1 | - | 2 | 196 | |||
BT019785.1 | - | 1 | 89 | |||
CR456505.1 | - | 2 | 204 | |||
CR541945.1 | - | 2 | 89 | |||
CR541968.1 | - | 2 | 89 | |||
GU187011.1 | - | 3 | 213 | |||
GU187012.1 | - | 2 | 826 | |||
JF323038.1 | - | 2 | 93 | |||
M23041.1 | - | 2 | 121 | |||
X06985.1 | - | 2 | 169 |