Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031679.2 | NP_001026849.1 | 7 | 1371 | ||
NM_001193460.1 | NP_001180389.1 | 8 | 1535 | |||
NM_001193461.1 | NP_001180390.1 | 7 | 1233 | |||
NM_198080.3 | NP_932346.1 | 6 | 1244 | |||
GenBank mRNA | AK299065.1 | - | 6 | 1184 | ||
AK316365.1 | - | 6 | 1216 | |||
AL833622.1 | - | 7 | 1288 | |||
AY358229.1 | - | 6 | 1179 | |||
BX640871.1 | - | 1 | 703 | |||
BX648776.1 | - | 7 | 1289 |