Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MITF SIRNA Assays ›
Gene Symbol:
MITF
Gene Name:
melanogenesis associated transcription factor
Gene Aliases:
CMM8, MI, WS2, WS2A, bHLHe32
Chromosome Location:
Chr.3: 69739435 - 69968337 on Build GRCh38
UniGene ID:
Hs.166017
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110564
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MITF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000248.3
NP_000239.1
9
1208
NM_001184967.1
NP_001171896.1
10
1421
NM_006722.2
NP_006713.1
10
1416
NM_198158.2
NP_937801.1
9
1190
NM_198159.2
NP_937802.1
10
1551
NM_198177.2
NP_937820.1
10
1407
NM_198178.2
NP_937821.2
9
1022
XM_005264754.1
XP_005264811.1
10
1541
XM_005264755.3
XP_005264812.1
10
1567
XM_006713164.2
XP_006713227.1
11
1561
XM_011533722.2
XP_011532024.1
10
1546
XM_011533723.2
XP_011532025.1
10
1566
XM_011533725.1
XP_011532027.1
10
1401
XM_011533726.1
XP_011532028.1
10
1383
XM_017006444.1
XP_016861933.1
10
9756
XM_017006445.1
XP_016861934.1
10
1528
XM_017006446.1
XP_016861935.1
10
1408
XM_017006447.1
XP_016861936.1
11
1659
XM_017006448.1
XP_016861937.1
11
1543
GenBank mRNA
AB006909.1
-
10
1504
AK291318.1
-
10
1523
AK296129.1
-
10
1421
AK297858.1
-
10
1353
AL110195.1
-
10
1507
BC026961.2
-
9
1120
3924101
BC065243.1
-
9
1165
6066096
GU355676.1
-
9
899
Z29678.1
-
9
1205
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
multicellular organism development
positive regulation of gene expression
protein sumoylation
melanocyte differentiation
regulation of apoptotic process
regulation of osteoclast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
mast cell migration
positive regulation of DNA-templated transcription, initiation
regulation of RNA biosynthetic process
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-gnqd7:80/10.239.161.169:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline