Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000139.4 | NP_000130.1 | 7 | 2341 | ||
NM_001256916.1 | NP_001243845.1 | 6 | 2206 | |||
XM_005273846.4 | XP_005273903.1 | 8 | 2656 | |||
XM_011544850.2 | XP_011543152.1 | 8 | 2274 |