Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000149.3 | NP_000140.1 | 3 | 1809 | ||
NM_001097639.1 | NP_001091108.1 | 3 | 1461 | |||
NM_001097640.1 | NP_001091109.1 | 3 | 1407 | |||
NM_001097641.1 | NP_001091110.1 | 2 | 1268 | |||
XM_011527865.1 | XP_011526167.1 | 3 | 1787 | |||
XM_011527866.1 | XP_011526168.1 | 3 | 1448 | |||
XM_011527867.1 | XP_011526169.1 | 3 | 1417 | |||
GenBank mRNA | AF131913.1 | - | 2 | 1340 | ||
AK291852.1 | - | 3 | 1436 | |||
BC108675.1 | - | 2 | 1265 | 5187745 | ||
U27326.1 | - | 3 | 1400 | |||
U27327.1 | - | 3 | 1454 | |||
U27328.1 | - | 4 | 1803 | |||
X53578.1 | - | 2 | 1243 |