Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000175.3 | NP_000166.2 | 3 | 394 | ||
NM_001184722.1 | NP_001171651.1 | 4 | 541 | |||
NM_001289789.1 | NP_001276718.1 | 4 | 541 | |||
NM_001289790.1 | NP_001276719.1 | 3 | 394 | |||
XM_005258764.2 | XP_005258821.1 | 4 | 360 | |||
XM_006723148.2 | XP_006723211.1 | 4 | 311 | |||
XM_011526754.2 | XP_011525056.1 | 5 | 776 | |||
GenBank mRNA | AB209575.1 | - | 2 | 1243 | ||
AK129884.1 | - | 4 | 325 | |||
AK130102.1 | - | 2 | 89 | |||
AK293446.1 | - | 3 | 270 | |||
AK294396.1 | - | 4 | 489 | |||
BC004982.1 | - | 3 | 262 | 2906270 | ||
DC299841.1 | - | 3 | 394 | |||
K03515.1 | - | 3 | 269 | |||
X16539.1 | - | 3 | 257 |