Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000373.3 | NP_000364.1 | 6 | 1736 | ||
NR_033434.1 | 5 | 1582 | ||||
NR_033437.1 | 7 | 1835 | ||||
GenBank mRNA | AB062285.1 | - | 6 | 1657 | ||
AK291306.1 | - | 6 | 1652 | |||
AL080099.2 | - | 5 | 1515 | |||
BC000364.2 | - | 6 | 1653 | 2964556 | ||
BC007511.2 | - | 6 | 1653 | 2964556 | ||
EU921886.1 | - | 6 | 1650 | |||
EU921887.1 | - | 5 | 1496 | |||
EU921888.1 | - | 5 | 1496 | |||
EU921889.1 | - | 6 | 1595 | |||
EU921890.1 | - | 6 | 1854 | |||
EU921891.1 | - | 7 | 1749 | |||
EU921892.1 | - | 6 | 1332 | |||
EU921893.1 | - | 6 | 1303 | |||
EU921895.1 | - | 3 | 1205 | |||
J03626.1 | - | 6 | 1734 |