Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF2RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000395.2 | NP_000386.1 | 8 | 1173 | ||
XM_005261340.3 | XP_005261397.1 | 8 | 1250 | |||
XM_011529903.2 | XP_011528205.1 | 8 | 1250 | |||
XM_011529904.2 | XP_011528206.1 | 8 | 1232 | |||
XM_011529905.2 | XP_011528207.1 | 8 | 1250 | |||
GenBank mRNA | AK302993.1 | - | 6 | 835 | ||
BC070085.1 | - | 8 | 1173 | 30344148 | ||
CR456428.1 | - | 7 | 1047 | |||
M59941.1 | - | 7 | 984 |