Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000416.2 | NP_000407.1 | 2 | 205 | ||
XM_006715470.2 | XP_006715533.1 | 3 | 566 | |||
XM_006715471.2 | XP_006715534.1 | 2 | 455 | |||
XM_011535793.2 | XP_011534095.1 | 3 | 1237 | |||
XM_011535794.1 | XP_011534096.1 | 3 | 182 | |||
XM_017010827.1 | XP_016866316.1 | 3 | 276 | |||
GenBank mRNA | AK222803.1 | - | 2 | 160 | ||
AK294252.1 | - | 3 | 248 | |||
AK297052.1 | - | 2 | 81 | |||
AK312674.1 | - | 2 | 159 | |||
BC005333.1 | - | 2 | 184 | 3950528 | ||
BT006814.1 | - | 2 | 102 | |||
DC349115.1 | - | 2 | 205 | |||
EF535103.1 | - | 2 | 145 | |||
J03143.1 | - | 2 | 150 | |||
KU178144.1 | - | 2 | 102 | |||
KU178145.1 | - | 2 | 102 |