Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000429.2 | NP_000420.1 | 6 | 887 | ||
XM_005269842.4 | XP_005269899.1 | 7 | 1117 | |||
XM_005269843.4 | XP_005269900.1 | 6 | 1006 | |||
GenBank mRNA | AK290820.1 | - | 6 | 753 | ||
BC018359.1 | - | 6 | 854 | 4341915 | ||
D49357.1 | - | 6 | 650 | |||
X69078.1 | - | 6 | 703 |