Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000429.2 | NP_000420.1 | 9 | 2034 | ||
XM_005269842.4 | XP_005269899.1 | 10 | 2264 | |||
XM_005269843.4 | XP_005269900.1 | 9 | 2153 | |||
GenBank mRNA | AK026931.1 | - | 3 | 1291 | ||
BC018359.1 | - | 9 | 2001 | 4341915 | ||
X69078.1 | - | 9 | 1850 |