Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002157.2 | NP_002148.1 | 2 | 533 | ||
GenBank mRNA | AK312104.1 | - | 2 | 129 | ||
BC023518.2 | - | 2 | 207 | 4304224 | ||
BG188453.1 | - | 1 | 94 | |||
CN264788.1 | - | 2 | 538 | |||
CR407688.1 | - | 1 | 93 | |||
U07550.1 | - | 2 | 134 | |||
X75821.1 | - | 1 | 93 |
Gene Symbol: HSPE1-MOB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202485.1 | NP_001189414.1 | 2 | 533 | ||
GenBank mRNA | BJ992310.1 | - | 2 | 151 | ||
CR990871.1 | - | 2 | 117 |