Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000628.4 | NP_000619.3 | 5, 6 | 744 | ||
GenBank mRNA | AK124057.1 | - | 4, 5 | 1055 | ||
AK299961.1 | - | 5, 6 | 645 | |||
AK313348.1 | - | 5, 6 | 737 | |||
BC001903.1 | - | 5, 6 | 723 | 3544611 | ||
BT009777.1 | - | 5, 6 | 638 | |||
Z17227.1 | - | 5, 6 | 681 |