Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000628.4 | NP_000619.3 | 7 | 1245 | ||
GenBank mRNA | AK124057.1 | - | 6 | 1556 | ||
AK299961.1 | - | 8 | 995 | |||
BC001903.1 | - | 7 | 1224 | 3544611 | ||
Z17227.1 | - | 7 | 1180 |