Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002161.5 | NP_002152.2 | 2 | 248 | ||
NM_013417.3 | NP_038203.2 | 2 | 418 | |||
NR_073446.1 | 2 | 248 | ||||
GenBank mRNA | AB209234.1 | - | 2 | 157 | ||
AK293014.1 | - | 2 | 175 | |||
AK301272.1 | - | 2 | 198 | |||
AK308202.1 | - | 2 | 182 | |||
BC065552.1 | - | 1 | 101 | 6043344 | ||
BX537417.1 | - | 2 | 186 | |||
BX537429.1 | - | 2 | 188 | |||
BX538109.1 | - | 2 | 188 | |||
BX647071.1 | - | 2 | 188 | |||
BX647236.1 | - | 2 | 180 | |||
BX647430.1 | - | 2 | 182 | |||
BX648567.1 | - | 2 | 188 | |||
D28473.1 | - | 2 | 336 | |||
DA335066.1 | - | 2 | 248 | |||
U04953.1 | - | 2 | 350 |