Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002164.5 | NP_002155.1 | 1, 2 | 209 | ||
GenBank mRNA | AK313259.1 | - | 1, 2 | 190 | ||
AU137351.1 | - | 1, 2 | 209 | |||
AY221100.1 | - | 1, 2 | 95 | |||
BC027882.1 | - | 1, 2 | 140 | 5208340 | ||
CD690354.1 | - | 1, 2 | 214 | |||
JF772862.1 | - | 1, 2 | 95 | |||
M34455.1 | - | 1, 2 | 734 | |||
X17668.1 | - | 1, 2 | 117 |