Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000755.3 | NP_000746.2 | 3, 4 | 695 | ||
NM_001257363.1 | NP_001244292.1 | 4, 5 | 794 | |||
XM_005251708.3 | XP_005251765.1 | 5, 6 | 857 | |||
XM_017014274.1 | XP_016869763.1 | 4, 5 | 862 | |||
XM_017014275.1 | XP_016869764.1 | 3, 4 | 515 | |||
GenBank mRNA | AK313931.1 | - | 4, 5 | 706 | ||
AL523652.3 | - | 3, 4 | 687 | |||
BC000723.2 | - | 3, 4 | 634 | 3504259 | ||
BI752361.1 | - | 3, 4 | 689 | |||
BT006801.1 | - | 3, 4 | 413 | |||
X78706.1 | - | 3, 4 | 408 |