Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000294.2 | NP_000285.1 | 10 | 1388 | ||
GenBank mRNA | AB209055.1 | - | 5 | 777 | ||
AK289492.1 | - | 10 | 1252 | |||
BC002541.2 | - | 10 | 1248 | 3138927 | ||
M31606.1 | - | 10 | 1271 |