Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080843.3 | NP_001074312.1 | 5 | 760 | ||
XM_005261715.4 | XP_005261772.1 | 5 | 738 | |||
GenBank mRNA | BC002415.1 | - | 5 | 765 | 3164017 | |
BC071700.1 | - | 5 | 732 | 4932043 | ||
BC146936.1 | - | 5 | 749 | 9056864 |
Gene Symbol: GSTT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000854.4 | NP_000845.2 | 5 | 880 | ||
NM_001302670.1 | NP_001289599.1 | 5 | 838 | |||
NR_126442.1 | 6 | 872 | ||||
NR_126445.1 | 5 | 880 | ||||
NR_126446.1 | 6 | 942 | ||||
GenBank mRNA | AK312043.1 | - | 5 | 774 | ||
BC146929.1 | - | 5 | 749 | 9056857 | ||
BC146940.1 | - | 5 | 850 | 9056868 | ||
BC146943.1 | - | 5 | 850 | 9056871 | ||
CD013993.1 | - | 6 | 750 | |||
CR456374.1 | - | 5 | 690 | |||
CR456500.1 | - | 5 | 730 | |||
CR456929.1 | - | 5 | 710 | |||
L38503.1 | - | 5 | 710 |