Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002167.4 | NP_002158.3 | 2 | 720 | ||
GenBank mRNA | A17546.1 | - | 2 | 475 | ||
AK225965.1 | - | 1 | 562 | |||
AK290003.1 | - | 2 | 457 | |||
BC003107.1 | - | 2 | 438 | 3543936 | ||
BM921819.1 | - | 751 | ||||
X66924.1 | - | 2 | 475 | |||
X69111.1 | - | 2 | 719 |