Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000876.3 | NP_000867.2 | 4, 5 | 658 | ||
GenBank mRNA | AB209668.1 | - | 3, 4 | 281 | ||
J03528.1 | - | 4, 5 | 657 | |||
Y00285.1 | - | 4, 5 | 657 |