Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204.6 | NP_001195.2 | 13 | 12000 | ||
XM_011511687.1 | XP_011509989.1 | 13 | 12014 | |||
GenBank mRNA | AK093832.1 | - | 1 | 2222 | ||
AK123579.1 | - | 2 | 2747 | |||
AY277593.1 | - | 1 | 3873 | |||
BC035097.1 | - | 1 | 2621 | |||
BC043650.2 | - | 1 | 3636 |