Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001824.4 | NP_001815.2 | 5 | 694 | ||
XM_017026240.1 | XP_016881729.1 | 4 | 579 | |||
GenBank mRNA | AK129878.1 | - | 5 | 592 | ||
AK300730.1 | - | 5 | 482 | |||
AK313281.1 | - | 5 | 592 | |||
BC007462.1 | - | 5 | 592 | 3934232 | ||
BT006793.1 | - | 4 | 519 | |||
M14780.1 | - | 5 | 597 |