Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000885.5 | NP_000876.3 | 7 | 1556 | ||
GenBank mRNA | AB208885.1 | - | 4 | 334 | ||
AK303220.1 | - | 3 | 231 | |||
AK303478.1 | - | 7 | 1043 | |||
BC016671.2 | - | 7 | 1021 | |||
L12002.1 | - | 7 | 1221 | |||
X16983.1 | - | 7 | 835 |