Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000885.5 | NP_000876.3 | 7 | 1560 | ||
GenBank mRNA | AB208885.1 | - | 4 | 338 | ||
AK303220.1 | - | 3 | 235 | |||
AK303478.1 | - | 7 | 1047 | |||
BC016671.2 | - | 7 | 1025 | |||
L12002.1 | - | 7 | 1225 | |||
X16983.1 | - | 7 | 839 |