Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000885.5 | NP_000876.3 | 14 | 2216 | ||
GenBank mRNA | AB208885.1 | - | 11 | 994 | ||
BC016671.2 | - | 14 | 1681 | |||
L12002.1 | - | 14 | 1881 | |||
X16983.1 | - | 14 | 1495 |