Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162407.1 | NP_001155879.1 | 6 | 1022 | ||
NM_004071.3 | NP_004062.2 | 6 | 742 | |||
NR_027855.1 | 5 | 651 | ||||
NR_027856.1 | 4 | 2142 | ||||
GenBank mRNA | AK025306.1 | - | 2 | 1317 | ||
AK289365.1 | - | 5 | 603 | |||
AK294295.1 | - | 6 | 1022 | |||
AK316127.1 | - | 6 | 713 | |||
BC028149.1 | - | 5 | 547 | |||
BC031549.1 | - | 6 | 638 | 4387232 | ||
L29219.1 | - | 6 | 716 | |||
L29222.1 | - | 5 | 625 | |||
M59287.1 | - | 5 | 626 |
Gene Symbol: PPIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK297804.1 | - | 11 | 1080 |