Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167928.1 | NP_001161400.1 | 5 | 527 | ||
NM_001167929.1 | NP_001161401.1 | 3 | 276 | |||
NM_001167930.1 | NP_001161402.1 | 3 | 276 | |||
NM_001167931.1 | NP_001161403.1 | 4 | 363 | |||
NM_002182.3 | NP_002173.1 | 4 | 363 | |||
NM_134470.3 | NP_608273.1 | 4 | 363 | |||
XM_017006347.1 | XP_016861836.1 | 3 | 276 | |||
GenBank mRNA | AB006537.1 | - | 4 | 312 | ||
AF029213.1 | - | 2 | 115 | |||
AF167343.1 | - | 2 | 106 | |||
AF538730.1 | - | 4 | 312 | |||
AF538731.1 | - | 3 | 225 | |||
AF538732.1 | - | 3 | 225 | |||
AF538733.1 | - | 3 | 225 | |||
AF538734.1 | - | 4 | 312 | |||
AK291669.1 | - | 5 | 522 | |||
BC053621.1 | - | 4 | 316 | 6160894 | ||
DA024801.1 | - | 3 | 278 | |||
EF591790.1 | - | 4 | 312 | |||
FJ998418.1 | - | 2 | 106 |