Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004760.2 | NP_004751.2 | 7 | 1154 | ||
XM_017012792.1 | XP_016868281.1 | 7 | 966 | |||
GenBank mRNA | AB011420.1 | - | 7 | 1120 | ||
BC023508.2 | - | 7 | 1110 | 3638004 | ||
BC047696.1 | - | 7 | 1123 | 5761182 |