Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002188.2 | NP_002179.2 | 3, 4 | 343 | ||
GenBank mRNA | AF043334.1 | - | 3, 4 | 290 | ||
BC096138.3 | - | 2, 3 | 1400 | 40002676 | ||
BC096139.1 | - | 3, 4 | 343 | 40002680 | ||
BC096140.3 | - | 2, 3 | 1398 | 40002684 | ||
BC096141.1 | - | 3, 4 | 343 | 40002686 | ||
L06801.1 | - | 3, 4 | 331 | |||
X69079.1 | - | 3, 4 | 343 |