Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018339.5 | NP_060809.3 | 4 | 2287 | ||
GenBank mRNA | AF038172.1 | - | 1 | 1157 | ||
BC007069.1 | - | 4 | 2181 | 3997678 |