Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001558.3 | NP_001549.2 | 7 | 3607 | ||
NR_026691.1 | 8 | 3737 | ||||
GenBank mRNA | AB209626.1 | - | 6 | 5108 | ||
BC028082.1 | - | 7 | 3584 | 5216986 | ||
U00672.1 | - | 7 | 3587 |