Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303233.1 | NP_001290162.1 | 2 | 234 | ||
NM_001303234.1 | NP_001290163.1 | 3 | 360 | |||
NM_001303236.1 | NP_001290165.1 | 2 | 606 | |||
NM_001303237.1 | NP_001290166.1 | 2 | 504 | |||
NM_002201.5 | NP_002192.2 | 2 | 504 | |||
XM_005254899.2 | XP_005254956.1 | 2 | 1767 | |||
XM_006720488.3 | XP_006720551.1 | 4 | 545 | |||
XM_017022147.1 | XP_016877636.1 | 2 | 1693 | |||
XM_017022148.1 | XP_016877637.1 | 3 | 383 | |||
GenBank mRNA | AK307769.1 | - | 2 | 302 | ||
BC007922.2 | - | 2 | 439 | 4309120 | ||
BC016341.1 | - | 2 | 307 | |||
BG759382.1 | - | 2 | 505 | |||
BI838264.1 | - | 3 | 313 | |||
BM691295.1 | - | 2 | 242 | |||
BT006952.1 | - | 1 | 146 | |||
BX339191.2 | - | 2 | 443 | |||
CN265233.1 | - | 2 | 294 | |||
CR456942.1 | - | 1 | 146 | |||
U88964.1 | - | 1 | 183 | |||
X89773.2 | - | 1 | 161 |