Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022766.5 | NP_073603.2 | 13 | 2102 | ||
XM_017028909.1 | XP_016884398.1 | 13 | 1899 | |||
GenBank mRNA | AB051433.1 | - | 12 | 1828 | ||
AB079066.1 | - | 13 | 2118 | |||
AK026892.1 | - | 1 | 260 | |||
AL832794.1 | - | 3 | 679 | |||
BC004278.1 | - | 3 | 679 | |||
BC108712.1 | - | 2 | 604 | |||
BU187541.1 | - | 2 | 604 |