Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002203.3 | NP_002194.2 | 2, 3 | 329 | ||
NR_073103.1 | 2, 3 | 329 | ||||
NR_073104.1 | 2, 3 | 329 | ||||
NR_073105.1 | 2, 3 | 329 | ||||
NR_073106.1 | 2, 3 | 329 | ||||
GenBank mRNA | AK307952.1 | - | 2, 3 | 318 | ||
BC143505.1 | - | 2, 3 | 215 | |||
BC143508.1 | - | 2, 3 | 215 | |||
BC143509.1 | - | 2, 3 | 215 | |||
BC143511.1 | - | 2, 3 | 215 | |||
BP280760.1 | - | 2, 3 | 331 | |||
X17033.1 | - | 2, 3 | 234 |