Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001628.2 | NP_001619.1 | 10 | 1193 | ||
XM_005250234.3 | XP_005250291.1 | 10 | 1202 | |||
GenBank mRNA | AB209796.1 | - | 9 | 1710 | ||
AF328729.1 | - | 10 | 1134 | |||
AL833410.1 | - | 1 | 2088 | |||
BC000260.1 | - | 10 | 1193 | 3357652 | ||
BC005387.1 | - | 10 | 1157 | 3960811 | ||
BC010391.1 | - | 10 | 1159 | 4049736 | ||
J04795.1 | - | 10 | 1128 | |||
J05017.1 | - | 10 | 1157 | |||
J05474.1 | - | 10 | 1151 | |||
M34720.1 | - | 10 | 1132 | |||
X15414.1 | - | 10 | 1164 |