Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001713.2 | NP_001704.2 | 8 | 2218 | ||
GenBank mRNA | BC012616.1 | - | 8 | 2194 | 4045543 | |
EU794592.1 | - | 8 | 2218 | |||
U50929.1 | - | 8 | 2139 |