Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002207.2 | NP_002198.2 | 2 | 268 | ||
GenBank mRNA | BC030198.1 | - | 2 | 475 | 5199296 | |
D25303.1 | - | 2 | 266 | |||
L24158.1 | - | 2 | 110 |