Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129889.2 | NP_001123361.1 | 1 | 536 | ||
NM_001922.4 | NP_001913.2 | 1 | 536 | |||
XM_017020401.1 | XP_016875890.1 | 1 | 539 | |||
GenBank mRNA | AJ000503.1 | - | 1 | 508 | ||
AK293115.1 | - | 1 | 319 | |||
BC028311.1 | - | 1 | 383 | 4705935 | ||
BP221841.1 | - | 1 | 537 | |||
D17547.1 | - | 1 | 523 | |||
DQ902581.1 | - | 1 | 401 | |||
L18967.1 | - | 1 | 493 | |||
S69231.1 | - | 1 | 458 |