Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052947.3 | NP_443179.3 | 4 | 2115 | ||
XM_011525801.2 | XP_011524103.1 | 4 | 2348 | |||
XM_011525802.2 | XP_011524104.1 | 2 | 2735 | |||
XM_017025535.1 | XP_016881024.1 | 4 | 2349 | |||
XM_017025536.1 | XP_016881025.1 | 4 | 2349 | |||
XM_017025537.1 | XP_016881026.1 | 4 | 2350 | |||
GenBank mRNA | AK092133.1 | - | 1 | 248 | ||
AK125243.1 | - | 1 | 1295 | |||
AL832018.2 | - | 1 | 462 | |||
AY044450.1 | - | 1 | 197 | |||
BX647639.1 | - | 1 | 981 | |||
BX647796.1 | - | 4 | 2117 |