Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052947.3 | NP_443179.3 | 5 | 3528 | ||
XM_011525801.2 | XP_011524103.1 | 5 | 3761 | |||
XM_011525802.2 | XP_011524104.1 | 3 | 4148 | |||
XM_017025535.1 | XP_016881024.1 | 5 | 3762 | |||
XM_017025536.1 | XP_016881025.1 | 5 | 3762 | |||
XM_017025537.1 | XP_016881026.1 | 5 | 3763 | |||
XM_017025538.1 | XP_016881027.1 | 2 | 2646 | |||
XM_017025539.1 | XP_016881028.1 | 2 | 1394 | |||
GenBank mRNA | AK092133.1 | - | 2 | 1646 | ||
AK092194.1 | - | 1 | 247 | |||
AL832018.2 | - | 2 | 1860 | |||
AY044450.1 | - | 2 | 1595 | |||
BX647446.1 | - | 1 | 1006 | |||
BX647639.1 | - | 2 | 2394 | |||
BX647796.1 | - | 5 | 3515 |