Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 14 | 1420 | ||
XM_006721162.1 | XP_006721225.1 | 13 | 1350 | |||
XM_011522928.1 | XP_011521230.1 | 14 | 1598 | |||
XM_011522929.1 | XP_011521231.1 | 14 | 1463 | |||
XM_011522930.2 | XP_011521232.1 | 14 | 1431 | |||
XM_011522931.2 | XP_011521233.1 | 14 | 1431 | |||
XM_017023012.1 | XP_016878501.1 | 14 | 1447 | |||
XM_017023013.1 | XP_016878502.1 | 11 | 1243 | |||
XM_017023014.1 | XP_016878503.1 | 9 | 1375 | |||
GenBank mRNA | AJ441184.1 | - | 14 | 1419 | ||
AK056456.1 | - | 14 | 1419 | |||
AK074168.1 | - | 12 | 1666 | |||
AK092025.1 | - | 6 | 575 | |||
AK096727.1 | - | 6 | 626 | |||
AK128387.1 | - | 14 | 1516 | |||
AK315590.1 | - | 14 | 1423 | |||
BC013735.1 | - | 12 | 1206 | |||
BC032542.1 | - | 14 | 1426 | 5534376 |