Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 24 | 3812 | ||
XM_006721162.1 | XP_006721225.1 | 23 | 3742 | |||
XM_011522928.1 | XP_011521230.1 | 24 | 3990 | |||
XM_011522929.1 | XP_011521231.1 | 24 | 3855 | |||
XM_017023012.1 | XP_016878501.1 | 24 | 3839 | |||
XM_017023013.1 | XP_016878502.1 | 21 | 3635 | |||
XM_017023014.1 | XP_016878503.1 | 19 | 3767 | |||
GenBank mRNA | AJ441184.1 | - | 24 | 3811 | ||
AK056456.1 | - | 24 | 3811 | |||
AK074168.1 | - | 22 | 4343 | |||
AK092025.1 | - | 16 | 2961 | |||
AK096727.1 | - | 16 | 3018 | |||
AK123470.1 | - | 3 | 2556 | |||
AK128387.1 | - | 24 | 3830 | |||
AY358187.1 | - | 2 | 1390 | |||
BC013735.1 | - | 22 | 3598 | |||
BC032542.1 | - | 24 | 3818 | 5534376 |