Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317958.1 | NP_001304887.1 | 8, 9 | 1327 | ||
NM_001317959.1 | NP_001304888.1 | 10, 11 | 1713 | |||
NM_152572.2 | NP_689785.1 | 8, 9 | 1320 | |||
XM_005272169.2 | XP_005272226.1 | 8, 9 | 859 | |||
XM_006716965.2 | XP_006717028.1 | 6, 7 | 632 | |||
XM_011518277.2 | XP_011516579.1 | 8, 9 | 859 | |||
XM_011518278.2 | XP_011516580.1 | 7, 8 | 700 | |||
XM_017014308.1 | XP_016869797.1 | 2, 3 | 382 | |||
GenBank mRNA | AK057266.1 | - | 8, 9 | 793 | ||
AK093446.1 | - | 8, 9 | 835 | |||
AK292186.1 | - | 8, 9 | 805 | |||
BC034776.1 | - | 8, 9 | 817 | 5175757 | ||
BC050576.1 | - | 8, 9 | 1320 | 5744517 |