Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCYL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020423.6 | NP_065156.5 | 12 | 1824 | ||
NM_181093.3 | NP_851607.2 | 13 | 1986 | |||
XM_006711465.1 | XP_006711528.1 | 13 | 1935 | |||
XM_011509801.1 | XP_011508103.1 | 13 | 2075 | |||
XM_011509802.1 | XP_011508104.1 | 13 | 1935 | |||
XM_011509803.1 | XP_011508105.1 | 12 | 1838 | |||
XM_017001862.1 | XP_016857351.1 | 12 | 1773 | |||
XM_017001863.1 | XP_016857352.1 | 12 | 1913 | |||
XM_017001864.1 | XP_016857353.1 | 12 | 1773 | |||
XM_017001865.1 | XP_016857354.1 | 12 | 1872 | |||
GenBank mRNA | AF540957.1 | - | 11 | 1654 | ||
AK292507.1 | - | 13 | 1932 | |||
AK304481.1 | - | 7 | 1416 | |||
AL117233.1 | - | 12 | 1707 | |||
AY144493.1 | - | 12 | 1816 | |||
BC014662.1 | - | 12 | 1761 | 4852719 | ||
BX647352.1 | - | 12 | 1913 |