Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL3RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002183.3 | NP_002174.1 | 3 | 429 | ||
XM_005274431.4 | XP_005274488.1 | 3 | 466 | |||
XM_005274432.1 | XP_005274489.1 | 3 | 259 | |||
XM_017029491.1 | XP_016884980.1 | 3 | 467 | |||
GenBank mRNA | AK290568.1 | - | 3 | 259 | ||
BC035407.1 | - | 3 | 429 | 5167281 | ||
M74782.1 | - | 3 | 226 |