Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002205.4 | NP_002196.4 | 2 | 314 | ||
GenBank mRNA | AK291570.1 | - | 2 | 259 | ||
AK311051.1 | - | 2 | 179 | |||
AK312414.1 | - | 2 | 314 | |||
AU132943.1 | - | 2 | 508 | |||
AU133210.1 | - | 2 | 314 | |||
BC008786.2 | - | 2 | 299 | 3629647 | ||
BM787112.1 | - | 2 | 314 | |||
M13918.2 | - | 2 | 201 | |||
X06256.1 | - | 2 | 269 |