Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002258.2 | NP_002249.1 | 4 | 409 | ||
GenBank mRNA | AK292022.1 | - | 4 | 424 | ||
BC113997.1 | - | 4 | 397 | |||
BC114516.1 | - | 4 | 397 | 40031215 | ||
BF432813.1 | - | 4 | 410 | |||
U11276.1 | - | 4 | 407 |