Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207024.1 | NP_001193953.1 | 5 | 841 | ||
NM_002355.3 | NP_002346.1 | 7 | 1099 | |||
XM_005253376.2 | XP_005253433.1 | 7 | 961 | |||
GenBank mRNA | AK222792.1 | - | 7 | 980 | ||
AK291143.1 | - | 7 | 945 | |||
AK312689.1 | - | 7 | 984 | |||
BC017111.1 | - | 5 | 499 | |||
BC024206.1 | - | 7 | 991 | 3530887 | ||
BM546062.1 | - | 2 | 254 | |||
M16985.1 | - | 7 | 969 |