Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002412.4 | NP_002403.2 | 5 | 830 | ||
XM_005252682.2 | XP_005252739.1 | 5 | 867 | |||
XM_017016275.1 | XP_016871764.1 | 4 | 653 | |||
GenBank mRNA | AK293593.1 | - | 5 | 800 | ||
BC000824.1 | - | 5 | 758 | 3454207 | ||
M29971.1 | - | 5 | 801 | |||
M31767.1 | - | 4 | 727 | |||
M60761.1 | - | 4 | 720 | |||
X54228.1 | - | 4 | 745 |